Assay Details
| Gene Symbol: LOC100912481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749683.2 | XP_003749731.1 | 1 - 2 | 47 | 120 | |
| XM_003749684.2 | XP_003749732.1 | 1 - 2 | 47 | 120 | ||
| XM_006236016.1 | XP_006236078.1 | 1 - 2 | 47 | 120 | ||
| XM_006236017.1 | XP_006236079.1 | 1 - 2 | 47 | 120 | ||
| XM_008762731.1 | XP_008760953.1 | 1 - 2 | 47 | 120 | ||
| Gene Symbol: Ewsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025632.2 | NP_001020803.2 | 1 - 2 | 45 | 120 | |
| XM_006251203.2 | XP_006251265.2 | 1 - 2 | 53 | 120 | ||
| XM_008770260.1 | XP_008768482.1 | 1 - 2 | 53 | 120 | ||
| GenBank mRNA | BC162004.1 | - | 1 - 2 | 45 | 9089248 | 120 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC162004.1,NM_001025632.2,XM_003749683.2,XM_003749684.2,XM_006236016.1,XM_006236017.1,XM_006251203.2,XM_008762731.1,XM_008770260.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100912481
Genomic Map