Assay Details
| Gene Symbol: LOC100912481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749683.2 | XP_003749731.1 | 4 - 5 | 261 | 104 | |
| XM_006236017.1 | XP_006236079.1 | 4 - 5 | 261 | 104 | ||
| XM_008762731.1 | XP_008760953.1 | 4 - 5 | 261 | 104 | ||
| Gene Symbol: Ewsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025632.2 | NP_001020803.2 | 4 - 5 | 259 | 104 | |
| XM_006251203.2 | XP_006251265.2 | 4 - 5 | 267 | 104 | ||
| XM_008770260.1 | XP_008768482.1 | 4 - 5 | 267 | 104 | ||
| GenBank mRNA | BC162004.1 | - | 4 - 5 | 259 | 9089248 | 104 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100912481
Genomic Map