Assay Details
| Gene Symbol: Mxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108364.1 | NP_001101834.1 | 3 - 4 | 402 | 78 | |
| XM_006251328.1 | XP_006251390.1 | 4 - 5 | 701 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mxd4
Genomic Map