Assay Details
| Gene Symbol: LOC102549464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770233.1 | XP_008768455.1 | 1 - 2 | 1705 | 89 | |
| Gene Symbol: Tbc1d14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034021.2 | NP_001029193.2 | 2 - 3 | 876 | 89 | |
| NM_001113365.1 | NP_001106836.1 | 3 - 4 | 1103 | 89 | ||
| XM_006251133.2 | XP_006251195.1 | 3 - 4 | 1109 | 89 | ||
| XM_006251134.2 | XP_006251196.1 | 2 - 3 | 801 | 89 | ||
| XM_008770323.1 | XP_008768545.1 | 3 - 4 | 1290 | 89 | ||
| GenBank mRNA | AB117752.1 | - | 3 - 4 | 1103 | 89 | |
| BC099760.1 | - | 2 - 3 | 849 | 7930485 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC102549464
Genomic Map