Assay Details
| Gene Symbol: Rnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007641.1 | NP_001007642.1 | 2 - 3 | 429 | 121 | |
| GenBank mRNA | AY461427.1 | - | 2 - 3 | 429 | 121 | |
| BC168142.1 | - | 457 | 9100645 | 121 | ||
| FQ210593.1 | - | 468 | 121 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rnd3
Genomic Map