Assay Details
| Gene Symbol: Lypd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234160.2 | XP_006234222.1 | 1 - 2 | 101 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006234160.2 transcripts.
Set Membership: |
|

Lypd6
Genomic Map