Assay Details
| Gene Symbol: Cd302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013916.1 | NP_001013938.1 | 3 - 4 | 526 | 90 | |
| GenBank mRNA | BC089829.1 | - | 3 - 4 | 526 | 7322853 | 90 |
| FQ209402.1 | - | 3 - 4 | 302 | 90 | ||
| FQ213169.1 | - | 3 - 4 | 320 | 90 | ||
| FQ215218.1 | - | 3 - 4 | 308 | 90 | ||
| FQ216383.1 | - | 3 - 4 | 308 | 90 | ||
| FQ219212.1 | - | 3 - 4 | 333 | 90 | ||
| FQ219651.1 | - | 3 - 4 | 306 | 90 | ||
| FQ219714.1 | - | 3 - 4 | 314 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd302
Genomic Map