Assay Details
| Gene Symbol: LOC679894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250641.2 | XP_006250703.1 | 6 - 7 | 6101 | 62 | |
| XM_006250642.2 | XP_006250704.1 | 7 - 8 | 6168 | 62 | ||
| XM_008770063.1 | XP_008768285.1 | 5 - 6 | 8597 | 62 | ||
| XM_008770064.1 | XP_008768286.1 | 4 - 5 | 8439 | 62 | ||
| XM_008770065.1 | XP_008768287.1 | 6 - 7 | 7420 | 62 | ||
| XM_008770066.1 | XP_008768288.1 | 6 - 7 | 8495 | 62 | ||
| XM_008770067.1 | XP_008768289.1 | 5 - 6 | 8337 | 62 | ||
| XM_008770068.1 | XP_008768290.1 | 6 - 7 | 7345 | 62 | ||
| XM_008770069.1 | XP_008768291.1 | 5 - 6 | 10601 | 62 | ||
More Information
Set Membership: |
|

LOC679894
Genomic Map