Assay Details
| Gene Symbol: LOC679894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250641.2 | XP_006250703.1 | 7 - 8 | 6224 | 72 | |
| XM_006250642.2 | XP_006250704.1 | 8 - 9 | 6291 | 72 | ||
| XM_006250643.2 | XP_006250705.1 | 6 - 7 | 6165 | 72 | ||
| XM_008770063.1 | XP_008768285.1 | 6 - 7 | 8720 | 72 | ||
| XM_008770064.1 | XP_008768286.1 | 5 - 6 | 8562 | 72 | ||
| XM_008770065.1 | XP_008768287.1 | 7 - 8 | 7543 | 72 | ||
| XM_008770066.1 | XP_008768288.1 | 7 - 8 | 8618 | 72 | ||
| XM_008770067.1 | XP_008768289.1 | 6 - 7 | 8460 | 72 | ||
| XM_008770068.1 | XP_008768290.1 | 7 - 8 | 7468 | 72 | ||
| XM_008770069.1 | XP_008768291.1 | 6 - 7 | 10724 | 72 | ||
More Information
Set Membership: |
|

LOC679894
Genomic Map