Assay Details
| Gene Symbol: Ibsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012587.2 | NP_036719.2 | 3 - 4 | 175 | 130 | |
| GenBank mRNA | BC127506.1 | - | 3 - 4 | 171 | 8369751 | 130 |
| DY314698.1 | - | 3 - 4 | 175 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ibsp
Genomic Map