Assay Details
| Gene Symbol: Ibsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012587.2 | NP_036719.2 | 1 - 2 | 59 | 82 | |
| GenBank mRNA | BC127506.1 | - | 1 - 2 | 55 | 8369751 | 82 |
| DY314698.1 | - | 1 - 2 | 59 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC127506.1,NM_012587.2 transcripts.
Set Membership: |
|

Ibsp
Genomic Map