Assay Details
| Gene Symbol: LOC679894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770069.1 | XP_008768291.1 | 1 - 2 | 2297 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008770069.1 transcripts.
Set Membership: |
|

LOC679894
Genomic Map