Assay Details
| Gene Symbol: Mustn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181368.3 | NP_852033.1 | 1 - 2 | 277 | 92 | |
| GenBank mRNA | AY254906.2 | - | 1 - 2 | 277 | 92 | |
| BC158570.1 | - | 1 - 2 | 292 | 9092791 | 92 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY254906.2,BC158570.1,NM_181368.3 transcripts.
Set Membership: |
|

Mustn1
Genomic Map