Assay Details
| Gene Symbol: Msmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019188.1 | NP_062061.1 | 2 - 3 | 133 | 75 | |
| XM_008770989.1 | XP_008769211.1 | 2 - 3 | 935 | 75 | ||
| GenBank mRNA | U65486.1 | - | 2 - 3 | 133 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msmb
Genomic Map