Assay Details
| Gene Symbol: Tnnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034105.1 | NP_001029277.1 | 2 - 3 | 54 | 68 | |
| XM_008770987.1 | XP_008769209.1 | 3 - 4 | 198 | 68 | ||
| GenBank mRNA | DQ062205.1 | - | 2 - 3 | 54 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnnc1
Genomic Map