Assay Details
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013134.2 | NP_001013152.2 | 2 - 3 | 497 | 77 | |
| XM_008770996.1 | XP_008769218.1 | 2 - 3 | 601 | 77 | ||
| GenBank mRNA | BC088323.1 | - | 2 - 3 | 509 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek4
Genomic Map