Assay Details
| Gene Symbol: Rbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191832.1 | NP_001178761.1 | 1 - 2 | 3120 | 118 | |
| GenBank mRNA | X69523.1 | - | 258 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbp3
Genomic Map