Assay Details
| Gene Symbol: Prl7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053364.1 | NP_445816.1 | 3 - 4 | 324 | 129 | |
| GenBank mRNA | AF139809.1 | - | 3 - 4 | 324 | 129 | |
| AF226609.1 | - | 3 - 4 | 296 | 129 | ||
| FQ219949.1 | - | 3 - 4 | 320 | 129 | ||
| FQ220120.1 | - | 3 - 4 | 271 | 129 | ||
More Information
Set Membership: |
|

Prl7d1
Genomic Map