Assay Details
| Gene Symbol: Ndufs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230731.1 | XP_006230793.1 | 1 - 2 | 254 | 139 | |
| GenBank mRNA | FQ213239.1 | - | 1 - 2 | 254 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006230731.1 transcripts.
Set Membership: |
|

Ndufs8
Genomic Map