Assay Details
| Gene Symbol: Scgb1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013051.1 | NP_037183.1 | 2 - 3 | 296 | 66 | |
| GenBank mRNA | BC069174.1 | - | 2 - 3 | 286 | 7096714 | 66 |
| J05536.1 | - | 2 - 3 | 296 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scgb1a1
Genomic Map