Assay Details
| Gene Symbol: Hnrnpul2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231006.2 | XP_006231068.1 | 9 - 10 | 1690 | 65 | |
| XM_219560.8 | XP_219560.5 | 9 - 10 | 1690 | 65 | ||
More Information
Set Membership: |
|

Hnrnpul2
Genomic Map