Assay Details
| Gene Symbol: Adrbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012776.1 | NP_036908.1 | 17 - 18 | 1799 | 77 | |
| GenBank mRNA | M87854.1 | - | 17 - 18 | 1799 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adrbk1
Genomic Map