Assay Details
| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024290.1 | NP_001019461.1 | 2 - 3 | 381 | 112 | |
| GenBank mRNA | BC094516.1 | - | 2 - 3 | 381 | 7314624 | 112 |
More Information
Set Membership: |
|

Gal3st3
Genomic Map