Assay Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107575.1 | NP_001101045.1 | 10 - 11 | 1127 | 84 | |
| XM_006230979.2 | XP_006231041.1 | 10 - 11 | 1346 | 84 | ||
| XM_006230981.2 | XP_006231043.1 | 10 - 11 | 1289 | 84 | ||
| XM_006230982.2 | XP_006231044.1 | 11 - 12 | 1216 | 84 | ||
| XM_006230983.2 | XP_006231045.1 | 10 - 11 | 1127 | 84 | ||
| XM_008760224.1 | XP_008758446.1 | 10 - 11 | 1598 | 84 | ||
| GenBank mRNA | BC168911.1 | - | 10 - 11 | 1242 | 7458553 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Taf6l
Genomic Map