Assay Details
Gene Symbol: Ms4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106336.1 | NP_001099806.1 | 1 - 2 | 59 | 75 | |
XM_006231032.2 | XP_006231094.1 | 1 - 2 | 336 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106336.1,XM_006231032.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
