Assay Details
Gene Symbol: Ms4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106336.1 | NP_001099806.1 | 5 - 6 | 564 | 94 | |
XM_006231032.2 | XP_006231094.1 | 5 - 6 | 841 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
