Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106321.2 | NP_001099791.2 | 19 - 20 | 4142 | 68 | |
| XM_006230726.2 | XP_006230788.1 | 19 - 20 | 4243 | 68 | ||
| GenBank mRNA | BC166786.1 | - | 19 - 20 | 4142 | 7935737 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Lrp5
Genomic Map