Assay Details
| Gene Symbol: Magec2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025713.1 | NP_001020884.1 | 3 - 4 | 197 | 76 | |
| XM_008773568.1 | XP_008771790.1 | 3 - 4 | 178 | 76 | ||
| GenBank mRNA | BC097464.1 | - | 3 - 4 | 197 | 7457620 | 76 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC097464.1, NM_001025713.1, XM_008773568.1 transcripts.
Set Membership: |
|

Magec2
Genomic Map