Assay Details
| Gene Symbol: LOC500124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024329.1 | NP_001019500.1 | 2 - 3 | 288 | 98 | |
| GenBank mRNA | BC078992.1 | - | 2 - 3 | 288 | 7112600 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC500124
Genomic Map