Assay Details
| Gene Symbol: Nt5c3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107862.1 | NP_001101332.1 | 3 - 4 | 311 | 126 | |
| XM_006236550.2 | XP_006236612.1 | 3 - 4 | 1459 | 126 | ||
| XM_006236552.2 | XP_006236614.1 | 2 - 3 | 353 | 126 | ||
| GenBank mRNA | BC166444.1 | - | 3 - 4 | 333 | 8363316 | 126 |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c3a
Genomic Map