Assay Details
| Gene Symbol: Nt5c3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107862.1 | NP_001101332.1 | 6 - 7 | 516 | 148 | |
| XM_006236550.2 | XP_006236612.1 | 6 - 7 | 1664 | 148 | ||
| XM_006236552.2 | XP_006236614.1 | 5 - 6 | 558 | 148 | ||
| GenBank mRNA | BC166444.1 | - | 6 - 7 | 538 | 8363316 | 148 |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c3a
Genomic Map