Assay Details
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729314.4 | XP_002729360.1 | 13 - 14 | 1974 | 64 | |
| XM_006236564.2 | XP_006236626.1 | 13 - 14 | 1976 | 64 | ||
| XM_006236565.2 | XP_006236627.1 | 13 - 14 | 1976 | 64 | ||
More Information
Set Membership: |
|

Avl9
Genomic Map