Assay Details
| Gene Symbol: Paqr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191077.1 | NP_001178006.1 | 3 - 4 | 244 | 87 | |
| XM_006232755.2 | XP_006232817.1 | 3 - 4 | 1246 | 87 | ||
| XM_008761210.1 | XP_008759432.1 | 3 - 4 | 1246 | 87 | ||
| XM_008761212.1 | XP_008759434.1 | 3 - 4 | 351 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Paqr6
Genomic Map