Assay Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047915.1 | NP_001041380.1 | 15 - 16 | 1708 | 90 | |
| XM_008765004.1 | XP_008763226.1 | 25 - 26 | 2776 | 90 | ||
| GenBank mRNA | AY390379.1 | - | 15 - 16 | 1708 | 90 | |
| FQ211964.1 | - | 17 - 18 | 1928 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P071820 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P458992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48954 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071820 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P458992 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P458992 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071820 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P458992 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P071820 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P458992 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73333 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97469 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99536 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69104 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58984 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23590 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10887794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887800 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887801 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887799 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887796 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887797 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887795 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887798 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10887774 | - | - |
| Rat Genome 230 2.0 Array | 1373666_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373666_at | - | - |
| Affymetrix HT Rat Focus Array | 1373666_at | - | - |
| Affymetrix HT Rat Focus Array | 1373666_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600012081.rn.1 | TC0600001699.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600012078.rn.1 | TC0600001699.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600012079.rn.1 | TC0600001699.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600012077.rn.1 | TC0600001699.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600012080.rn.1 | TC0600001699.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373666_PM_at | - | - |
| Rat Expression Set 230 | 1373666_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10887774 | - | - |
| Rat Genome U34 Set | rc_AI102560_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21262508 | - | - |

Rapgef5
Genomic Map