Assay Details
| Gene Symbol: Sema4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106902.1 | NP_001100372.1 | 6 - 7 | 1215 | 57 | |
| XM_006244758.2 | XP_006244820.1 | 4 - 5 | 2415 | 57 | ||
| XM_006244759.2 | XP_006244821.1 | 5 - 6 | 910 | 57 | ||
| XM_006244760.2 | XP_006244822.1 | 5 - 6 | 2035 | 57 | ||
| XM_008766995.1 | XP_008765217.1 | 5 - 6 | 907 | 57 | ||
| XM_008766996.1 | XP_008765218.1 | 5 - 6 | 4520 | 57 | ||
| XM_008766997.1 | XP_008765219.1 | 5 - 6 | 2038 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4c
Genomic Map