Assay Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030844.2 | NP_110471.2 | 7 - 8 | 874 | 108 | |
| XM_006236092.2 | XP_006236154.1 | 7 - 8 | 970 | 108 | ||
| XM_006236093.2 | XP_006236155.1 | 7 - 8 | 1192 | 108 | ||
| XM_006236094.2 | XP_006236156.1 | 6 - 7 | 820 | 108 | ||
| XM_006236095.2 | XP_006236157.1 | 6 - 7 | 806 | 108 | ||
| XM_008762728.1 | XP_008760950.1 | 7 - 8 | 970 | 108 | ||
| GenBank mRNA | BC063810.1 | - | 7 - 8 | 893 | 108 | |
| BC078812.1 | - | 7 - 8 | 874 | 7112043 | 108 | |
| L20900.1 | - | 7 - 8 | 1080 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ica1
Genomic Map