Assay Details
| Gene Symbol: Cdc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013103.2 | NP_001013121.2 | 4 - 5 | 516 | 130 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of cAMP-mediated signaling
response to growth factor
protein K48-linked ubiquitination
positive regulation of inclusion body assembly
molecular_function
ubiquitin-protein transferase activity
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
protein polyubiquitination
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of cAMP-mediated signaling
response to growth factor
protein K48-linked ubiquitination
positive regulation of inclusion body assembly
molecular_function
ubiquitin-protein transferase activity
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P152236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1112 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P152236 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P152236 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P152236 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P491878 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13979 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10900823 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900830 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900826 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900829 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900828 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811247 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900824 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900822 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900825 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900827 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10900821 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10811246 | - | - |
| Rat Genome 230 2.0 Array | 1388286_a_at | - | - |
| Rat Genome 230 2.0 Array | 1388286_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1388400_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371278_at | - | - |
| Rat Genome 230 2.0 Array | 1388400_at | - | - |
| Rat Genome 230 2.0 Array | 1371278_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388400_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388286_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1371278_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388400_at | - | - |
| Affymetrix HT Rat Focus Array | 1388286_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1371278_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700017721.rn.1 | TC0700002207.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700017725.rn.1 | TC0700002207.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700017722.rn.1 | TC0700002207.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700017723.rn.1 | TC0700002207.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388400_PM_at | - | - |
| Rat Expression Set 230 | 1388286_a_at | - | - |
| Rat Expression Set 230 | 1371278_PM_at | - | - |
| Rat Expression Set 230 | 1371278_at | - | - |
| Rat Expression Set 230 | 1388400_at | - | - |
| Rat Expression Set 230 | 1388286_PM_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900821 | - | - |
| Rat Genome U34 Set | rc_AA799788_s_at | - | - |
| Rat Genome U34 Set | L38482_g_at | - | - |
| Rat Genome U34 Set | L38482_at | - | - |
| Rat Genome U34 Set | rc_AA818770_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22122182 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530485 | - | - |
| RatRef-12 Expression BeadChip | ILMN_161578 | - | - |

Cdc34
Genomic Map