Assay Details
| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047100.2 | NP_001040565.2 | 11 - 12 | 1921 | 82 | |
| XM_006240931.1 | XP_006240993.1 | 13 - 14 | 2525 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss9
Genomic Map