Assay Details
| Gene Symbol: Madcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019317.1 | NP_062190.1 | 1 - 2 | 72 | 93 | |
| XM_008765179.1 | XP_008763401.1 | 1 - 2 | 384 | 93 | ||
| GenBank mRNA | D87840.1 | - | 1 - 2 | 72 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D87840.1, NM_019317.1, XM_008765179.1 transcripts.
Set Membership: |
|

Madcam1
Genomic Map