Assay Details
| Gene Symbol: Madcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019317.1 | NP_062190.1 | 3 - 4 | 662 | 101 | |
| XM_008765179.1 | XP_008763401.1 | 3 - 4 | 974 | 101 | ||
| GenBank mRNA | D87840.1 | - | 3 - 4 | 662 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Madcam1
Genomic Map