Assay Details
Gene Symbol: Dohh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025006.1 | NP_001020177.1 | 2 - 3 | 395 | 123 | |
XM_006240937.2 | XP_006240999.1 | 2 - 3 | 497 | 123 | ||
XM_006240938.2 | XP_006241000.1 | 3 - 4 | 687 | 123 | ||
XM_006240939.2 | XP_006241001.1 | 3 - 4 | 684 | 123 | ||
GenBank mRNA | BC087658.1 | - | 2 - 3 | 395 | 7314221 | 123 |
FQ228684.1 | - | 2 - 3 | 461 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
