Assay Details
| Gene Symbol: Fam195b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108311.1 | NP_001101781.1 | 1 - 2 | 54 | 76 | |
| XM_006247896.2 | XP_006247958.1 | 2 - 3 | 85 | 76 | ||
| XM_008768479.1 | XP_008766701.1 | 2 - 3 | 881 | 76 | ||
| GenBank mRNA | BC098826.1 | - | 1 - 2 | 61 | 76 | |
| BC158709.1 | - | 1 - 2 | 55 | 9034656 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC158709.1,NM_001108311.1,XM_006247896.2,XM_008768479.1 transcripts.
This assay is in the 5'UTR region of the BC158709.1,NM_001108311.1,XM_006247896.2,XM_008768479.1 transcripts.
Set Membership: |
|

Fam195b
Genomic Map