Assay Details
| Gene Symbol: Fam195b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108311.1 | NP_001101781.1 | 2 - 3 | 172 | 65 | |
| XM_006247896.2 | XP_006247958.1 | 3 - 4 | 203 | 65 | ||
| XM_008768479.1 | XP_008766701.1 | 3 - 4 | 999 | 65 | ||
| GenBank mRNA | BC098826.1 | - | 2 - 3 | 179 | 65 | |
| BC158709.1 | - | 2 - 3 | 173 | 9034656 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam195b
Genomic Map