Assay Details
| Gene Symbol: Tsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108268.1 | NP_001101738.1 | 2 - 3 | 671 | 100 | |
| XM_006245996.2 | XP_006246058.1 | 3 - 4 | 582 | 100 | ||
| XM_006245997.2 | XP_006246059.1 | 3 - 4 | 561 | 100 | ||
| GenBank mRNA | BC167035.1 | - | 1 - 2 | 799 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsr3
Genomic Map