Assay Details
Gene Symbol: Mettl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008283.1 | NP_001008284.1 | 2 - 3 | 267 | 63 | |
XM_006247798.1 | XP_006247860.1 | 2 - 3 | 253 | 63 | ||
GenBank mRNA | BC086594.1 | - | 2 - 3 | 267 | 7310322 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
