Assay Details
| Gene Symbol: Mettl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008283.1 | NP_001008284.1 | 3 - 4 | 501 | 108 | |
| XM_006247798.1 | XP_006247860.1 | 3 - 4 | 487 | 108 | ||
| GenBank mRNA | BC086594.1 | - | 3 - 4 | 501 | 7310322 | 108 |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl23
Genomic Map