Assay Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107223.1 | NP_001100693.1 | 2 - 3 | 260 | 83 | |
| XM_006251092.2 | XP_006251154.1 | 2 - 3 | 286 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb49
Genomic Map