Assay Details
| Gene Symbol: Asmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144759.2 | NP_653360.2 | 4 - 5 | 463 | 106 | |
| XM_006248991.2 | XP_006249053.1 | 4 - 5 | 1249 | 106 | ||
| XM_006248992.1 | XP_006249054.1 | 3 - 4 | 294 | 106 | ||
| GenBank mRNA | EU741665.1 | - | 4 - 5 | 443 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asmt
Genomic Map