Assay Details
| Gene Symbol: Pdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022595.1 | NP_072117.1 | 4 - 5 | 361 | 71 | |
| GenBank mRNA | BC088140.1 | - | 4 - 5 | 375 | 7382664 | 71 |
| U26541.1 | - | 4 - 5 | 361 | 71 | ||
| U41744.1 | - | 4 - 5 | 382 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdap1
Genomic Map