Assay Details
| Gene Symbol: Bud31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053556.1 | NP_446008.1 | 3 - 4 | 396 | 107 | |
| XM_006248887.1 | XP_006248949.1 | 3 - 4 | 513 | 107 | ||
| XM_006248889.1 | XP_006248951.1 | 4 - 5 | 381 | 107 | ||
| XM_008769003.1 | XP_008767225.1 | 4 - 5 | 610 | 107 | ||
| GenBank mRNA | AF058791.1 | - | 3 - 4 | 396 | 107 | |
| BC058456.1 | - | 4 - 5 | 423 | 6887068 | 107 | |
| FQ230665.1 | - | 1 - 2 | 124 | 107 | ||
| FQ232789.1 | - | 1 - 2 | 124 | 107 | ||
| FQ232851.1 | - | 1 - 2 | 124 | 107 | ||
| FQ232911.1 | - | 1 - 2 | 124 | 107 | ||
| FQ235300.1 | - | 1 - 2 | 124 | 107 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bud31
Genomic Map